Search
Search

| Assay Name: | hsa-miR-374b-3p |
|---|---|
| miRBase Accession Number: | MI0005566 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUUAGCAGGUUGUAUUAUCAUU |
| Species: | Human, Pteropus alecto, Rabbit |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | CUUAGCAGGUUGUAUUAUCAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374b-3p | MIMAT0004956 |
| Pteropus alecto | pal-miR-374b-3p | MIMAT0039990 |
| Rabbit | ocu-miR-374b-3p | MIMAT0048409 |