Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12855
          Assay Name: hsa-let-7e-3p
          miRBase Accession Number: MI0000066
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 51692786 - 51692864 [+] on Build GRCh38
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Species: Human, Mouse, Rat, Cricetulus griseus, Goat, Guinea pig, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12855

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUAUACGGCCUCCUAGCUUUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7e-3p MIMAT0004485
          Mouse mmu-let-7e-3p MIMAT0017016
          Rat rno-let-7e-3p MIMAT0004706
          Cricetulus griseus cgr-let-7e MIMAT0041132
          Goat chi-let-7e-3p MIMAT0035889
          Guinea pig cpo-let-7e-3p MIMAT0046827
          View More Rows
          Rhesus monkey mml-let-7e-3p MIMAT0026792

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7e
          Stem-loop Accession # MI0000066
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location Chr. 19 - 51692786 - 51692864 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004485
          miRBase ID: hsa-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 19 - 51692786 - 51692864 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7e
          Stem-loop Accession # MI0000561
          Stem-loop Sequence
          CGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCC
          Chromosome Location Chr. 17 - 17830352 - 17830444 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017016
          miRBase ID: mmu-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 17 - 17830352 - 17830444 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7e
          Stem-loop Accession # MI0000832
          Stem-loop Sequence
          CGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCC
          Chromosome Location Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004706
          miRBase ID: rno-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: Chr. 1 - 59704381 - 59704473 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-let-7e
          Stem-loop Accession # MI0033060
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAAGACACCCGAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041132
          miRBase ID: cgr-let-7e
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7e
          Stem-loop Accession # MI0030583
          Stem-loop Sequence
          ACCUGCCGCGCCCCCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGGCUGCGCCCUGCACGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035889
          miRBase ID: chi-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7e
          Stem-loop Accession # MI0038546
          Stem-loop Sequence
          UGAGGUAGGAGGUUGUAUAGUUGAGAAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046827
          miRBase ID: cpo-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7e
          Stem-loop Accession # MI0007576
          Stem-loop Sequence
          CCCGGGCUGAGGUAGGAGGUUGUAUAGUUGAGGAGGACACCCAAGGAGAUCACUAUACGGCCUCCUAGCUUUCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026792
          miRBase ID: mml-let-7e-3p
          Mature miRNA Sequence: CUAUACGGCCUCCUAGCUUUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295173_pri
          TaqMan™ Pri-miRNA Assay : Rn03465947_pri
          TaqMan™ Pri-miRNA Assay : Mm03306232_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12855
          Ambion® Pre-miR™ miRNA Precursor : PM12855
          TaqMan™ MicroRNA Assay : 002407
          mirVana® miRNA mimic : MC12855
          TaqMan™ Advanced miRNA Assay : rno479281_mir
          TaqMan™ Advanced miRNA Assay : 479281_mir
          TaqMan™ Advanced miRNA Assay : mmu483017_mir


          miRBase Alias:

          Human: hsa-let-7e* (v17)
          Mouse: mmu-let-7e* (v17)
          Rat: rno-let-7e* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline