Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12857
          Assay Name: hsa-miR-200b-5p
          miRBase Accession Number: MI0000342
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 1167104 - 1167198 [+] on Build GRCh38
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Species: Human, Mouse, Rat, Chrysemys picta, Columba livia, Dasypus novemcinctus, Dog, Guinea pig
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12857

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          CAUCUUACUGGGCAGCAUUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200b-5p MIMAT0004571
          Mouse mmu-miR-200b-5p MIMAT0004545
          Rat rno-miR-200b-5p MIMAT0017152
          Chrysemys picta cpi-miR-200b-5p MIMAT0037822
          Columba livia cli-miR-200b-5p MIMAT0038577
          Dasypus novemcinctus dno-miR-200b-5p MIMAT0047740
          View More Rows
          Dog cfa-miR-200b MIMAT0009864
          Guinea pig cpo-miR-200b-5p MIMAT0047091

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200b
          Stem-loop Accession # MI0000342
          Stem-loop Sequence
          CCAGCUCGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUGGAGUCAGGUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACG
          Chromosome Location Chr. 1 - 1167104 - 1167198 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004571
          miRBase ID: hsa-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: Chr. 1 - 1167104 - 1167198 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200b
          Stem-loop Accession # MI0000243
          Stem-loop Sequence
          GCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGC
          Chromosome Location Chr. 4 - 156055681 - 156055750 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004545
          miRBase ID: mmu-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: Chr. 4 - 156055681 - 156055750 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-200b
          Stem-loop Accession # MI0000944
          Stem-loop Sequence
          CCAACUUGGGCAGCCGUGGCCAUCUUACUGGGCAGCAUUGGAUAGUGUCUGAUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGAGCCCUGCACG
          Chromosome Location Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017152
          miRBase ID: rno-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: Chr. 5 - 173490144 - 173490238 [-] on Build Rnor_6.0

          Chrysemys picta

          Stem-loop ID cpi-mir-200b
          Stem-loop Accession # MI0029525
          Stem-loop Sequence
          ACCCUGGGAUGCCAUUACCAUCUUACUGGGCAGCAUUGGAUGUUUUCUGUGUUUCUAAUACUGCCUGGUAAUGAUGAUUAUGGUGUCUCGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037822
          miRBase ID: cpi-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-200b
          Stem-loop Accession # MI0030006
          Stem-loop Sequence
          CCUGAGAUGCCAUUACCAUCUUACUGGGCAGCAUUGGAUGUUUUCUGUCUUUCUAAUACUGCCUGGUAAUGAUGAUUAUGGUGUUUCGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038577
          miRBase ID: cli-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-200b
          Stem-loop Accession # MI0039034
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAACGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047740
          miRBase ID: dno-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-200b
          Stem-loop Accession # MI0010361
          Stem-loop Sequence
          GCCAUCGCCAUCUUACUGGGCAGCAUUGGAUGGUGUCAGGUCUCUAAUACUGCCUGGUAAUGAUGACGGCGGGGCCCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009864
          miRBase ID: cfa-miR-200b
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200b
          Stem-loop Accession # MI0038693
          Stem-loop Sequence
          CAUCUUACUGGGCAGCAUUGGAUGGUGUCUGGUCUCUAAUACUGCCUGGUAAUGAUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047091
          miRBase ID: cpo-miR-200b-5p
          Mature miRNA Sequence: CAUCUUACUGGGCAGCAUUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464804_pri
          TaqMan™ Pri-miRNA Assay : Hs03303027_pri
          TaqMan™ Pri-miRNA Assay : Mm03306509_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12857
          Ambion® Pre-miR™ miRNA Precursor : PM12857
          TaqMan™ MicroRNA Assay : 002274
          mirVana® miRNA mimic : MC12857
          TaqMan™ Advanced miRNA Assay : 478753_mir
          TaqMan™ Advanced miRNA Assay : mmu478753_mir
          TaqMan™ Advanced miRNA Assay : rno478753_mir


          miRBase Alias:

          Human: hsa-miR-200b* (v17)
          Mouse: mmu-miR-200b* (v17)
          Rat: rno-miR-200b* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline