Search
Search

| Assay Name: | mmu-miR-101a-5p |
|---|---|
| miRBase Accession Number: | MI0000148 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAGUUAUCACAGUGCUGAUGC |
| Species: | Mouse, Rat, Cricetulus griseus, Goat, Oryzias latipes, Taeniopygia guttata |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UCAGUUAUCACAGUGCUGAUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-101a-5p | MIMAT0004526 |
| Rat | rno-miR-101a-5p | MIMAT0004726 |
| Cricetulus griseus | cgr-miR-101a | MIMAT0041145 |
| Goat | chi-miR-101-5p | MIMAT0035899 |
| Oryzias latipes | ola-miR-101a-5p | MIMAT0022542 |
| Taeniopygia guttata | tgu-miR-101-2-5p | MIMAT0014631 |