Search
Search

| Assay Name: | hsa-miR-127-5p |
|---|---|
| miRBase Accession Number: | MI0000472 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGAAGCUCAGAGGGCUCUGAU |
| Species: | Human, Mouse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000080, mir-127 |
Mature miRNA Sequence: | CUGAAGCUCAGAGGGCUCUGAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-127-5p | MIMAT0004604 |
| Mouse | mmu-miR-127-5p | MIMAT0004530 |