Search
Search

| Assay Name: | hsa-miR-1202 |
|---|---|
| miRBase Accession Number: | MI0006334 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGCCAGCUGCAGUGGGGGAG |
| Species: | Human, Pan troglodytes, Pongo pygmaeus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000613, mir-1202 |
Mature miRNA Sequence: | GUGCCAGCUGCAGUGGGGGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1202 | MIMAT0005865 |
| Pan troglodytes | ptr-miR-1202 | MIMAT0007956 |
| Pongo pygmaeus | ppy-miR-1202 | MIMAT0016152 |