Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH14066
          Assay Name: hsa-miR-137-5p
          miRBase Accession Number: MI0000454
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 98046070 - 98046171 [-] on Build GRCh38
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Species: Human, Mouse, Columba livia, Cricetulus griseus, Ornithorhynchus anatinus, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH14066

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000106, mir-137

          Mature miRNA Sequence:

          ACGGGUAUUCUUGGGUGGAUAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-137-5p MIMAT0037310
          Mouse mmu-miR-137-5p MIMAT0016986
          Columba livia cli-miR-137a-5p MIMAT0038518
          Cricetulus griseus cgr-miR-137-5p MIMAT0023762
          Ornithorhynchus anatinus oan-miR-137a-5p MIMAT0006837
          Taeniopygia guttata tgu-miR-137-5p MIMAT0026995

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-137
          Stem-loop Accession # MI0000454
          Stem-loop Sequence
          GGUCCUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACCAGCGGCA
          Chromosome Location Chr. 1 - 98046070 - 98046171 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0037310
          miRBase ID: hsa-miR-137-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: Chr. 1 - 98046070 - 98046171 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-137
          Stem-loop Accession # MI0000163
          Stem-loop Sequence
          CUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGG
          Chromosome Location Chr. 3 - 118433857 - 118433929 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0016986
          miRBase ID: mmu-miR-137-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: Chr. 3 - 118433857 - 118433929 [+] on Build GRCm38

          Columba livia

          Stem-loop ID cli-mir-137a
          Stem-loop Accession # MI0029971
          Stem-loop Sequence
          CUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038518
          miRBase ID: cli-miR-137a-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-137
          Stem-loop Accession # MI0020399
          Stem-loop Sequence
          CUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023762
          miRBase ID: cgr-miR-137-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-137a
          Stem-loop Accession # MI0006687
          Stem-loop Sequence
          GCUUCGUCUUUCUGACUCUCUUCGGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAGUAUCUGCGGCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006837
          miRBase ID: oan-miR-137a-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-137
          Stem-loop Accession # MI0013757
          Stem-loop Sequence
          GGUGACGGGUAUUCUUGGGUGGAUAAUACGGAUUACGUUGUUAUUGCUUAAGAAUACGCGUAGUCGAGGAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026995
          miRBase ID: tgu-miR-137-5p
          Mature miRNA Sequence: ACGGGUAUUCUUGGGUGGAUAAU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306305_pri
          TaqMan™ Pri-miRNA Assay : Hs03303136_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM14066
          Ambion® Pre-miR™ miRNA Precursor : PM14066
          TaqMan™ MicroRNA Assay : 006016_mat
          mirVana® miRNA mimic : MC14066
          TaqMan™ Advanced miRNA Assay : 483102_mir
          TaqMan™ Advanced miRNA Assay : mmu481541_mir


          miRBase Alias:

          Mouse: mmu-miR-137* (v17)
          Ochrobactrum anthropi: oan-miR-137a* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-crr4t:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0