Search
Search

| Assay Name: | bkv-miR-B1-3p |
|---|---|
| miRBase Accession Number: | MI0009981 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCUUGAUCCAUGUCCAGAGUC |
| Species: | BK polyomavirus, JC polyomavirus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000637, mir-B1 |
Mature miRNA Sequence: | UGCUUGAUCCAUGUCCAGAGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| BK polyomavirus | bkv-miR-B1-3p | MIMAT0009150 |
| JC polyomavirus | jcv-miR-J1-3p | MIMAT0009148 |