Search
Search

| Assay Name: | mdo-miR-150-5p |
|---|---|
| miRBase Accession Number: | MI0012504 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUCCCAACCCUUGUACCAGAGU |
| Species: | Monodelphis domestica |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000197, mir-150 |
Mature miRNA Sequence: | UCUCCCAACCCUUGUACCAGAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-150-5p | MIMAT0012748 |