Search
Search

| Assay Name: | eca-miR-615-3p |
|---|---|
| miRBase Accession Number: | MI0012729 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCUC |
| Species: | Horse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Horse | eca-miR-615-3p | MIMAT0012977 |