Search
Search

| Assay Name: | mdo-miR-28 |
|---|---|
| miRBase Accession Number: | MI0012516 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAGGAGCUCACAGUCGAAUG |
| Species: | Monodelphis domestica |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCGAAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-28 | MIMAT0012759 |