Search
Search

| Assay Name: | hsa-miR-3146 |
|---|---|
| miRBase Accession Number: | MI0014172 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUGCUAGGAUAGAAAGAAUGG |
| Species: | Human |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001626, mir-3146 |
Mature miRNA Sequence: | CAUGCUAGGAUAGAAAGAAUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3146 | MIMAT0015018 |