Search
Search

| Assay Name: | aae-miR-34-3p |
|---|---|
| miRBase Accession Number: | MI0013456 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAACCACUAUCCGCCCUGCCGCC |
| Species: | Aedes aegypti |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000039, mir-34 |
Mature miRNA Sequence: | CAACCACUAUCCGCCCUGCCGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aedes aegypti | aae-miR-34-3p | MIMAT0014246 |