Search
Search

| Assay Name: | mmu-miR-138-2-3p |
|---|---|
| miRBase Accession Number: | MI0000164 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCUAUUUCACGACACCAGGGU |
| Species: | Mouse, Rat, Guinea pig, Rabbit |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000075, mir-138 |
Mature miRNA Sequence: | GCUAUUUCACGACACCAGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-138-2-3p | MIMAT0016987 |
| Rat | rno-miR-138-2-3p | MIMAT0017127 |
| Guinea pig | cpo-miR-138-2-3p | MIMAT0047001 |
| Rabbit | ocu-miR-138-2-3p | MIMAT0048238 |