Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH19615
          Assay Name: hsa-miR-3913-5p
          miRBase Accession Number: MI0016417
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 69584722 - 69584823 [-] on Build GRCh38
          Mature miRNA Sequence: UUUGGGACUGAUCUUGAUGUCU
          Species: Human
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH19615

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001134, mir-3913

          Mature miRNA Sequence:

          UUUGGGACUGAUCUUGAUGUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3913-5p MIMAT0018187

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3913-1
          Stem-loop Accession # MI0016417
          Stem-loop Sequence
          UUGUUUAUAAUAAACUGAAAUAUUUGGGACUGAUCUUGAUGUCUGCCAAAACCUUGGCAGACAUCAAGAUCAGUCCCAAAUAUUUCAGUUUAUUAUAGACAG
          Chromosome Location Chr. 12 - 69584722 - 69584823 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0018187
          miRBase ID: hsa-miR-3913-5p
          Mature miRNA Sequence: UUUGGGACUGAUCUUGAUGUCU
          Chromosome Location: Chr. 12 - 69584722 - 69584823 [-] on Build GRCh38

          Stem-loop ID hsa-mir-3913-2
          Stem-loop Accession # MI0016418
          Stem-loop Sequence
          UGUCUAUAAUAAACUGAAAUAUUUGGGACUGAUCUUGAUGUCUGCCAAGGUUUUGGCAGACAUCAAGAUCAGUCCCAAAUAUUUCAGUUUAUUAUAAACA
          Chromosome Location Chr. 12 - 69584723 - 69584822 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0018187
          miRBase ID: hsa-miR-3913-5p
          Mature miRNA Sequence: UUUGGGACUGAUCUUGAUGUCU
          Chromosome Location: Chr. 12 - 69584722 - 69584823 [-] on Build GRCh38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04226450_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19615
          Ambion® Pre-miR™ miRNA Precursor : PM19615
          TaqMan™ MicroRNA Assay : 462679_mat
          mirVana® miRNA mimic : MC19615
          TaqMan™ Advanced miRNA Assay : 479735_mir


          miRBase Alias:

          Human: hsa-miR-3913 (v16)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0