Search
Search

| Assay Name: | mmu-miR-448-5p |
|---|---|
| miRBase Accession Number: | MI0001638 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAACAUCCUGCAUAGUGCUGCC |
| Species: | Mouse, Rabbit |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000149, mir-448 |
Mature miRNA Sequence: | GAACAUCCUGCAUAGUGCUGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-448-5p | MIMAT0017176 |
| Rabbit | ocu-miR-448-5p | MIMAT0048451 |