Search
Search

| Assay Name: | rno-miR-127-5p |
|---|---|
| miRBase Accession Number: | MI0000899 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGAAGCUCAGAGGGCUCUGAUU |
| Species: | Rat, Dasypus novemcinctus, Guinea pig, Rabbit |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000080, mir-127 |
Mature miRNA Sequence: | CUGAAGCUCAGAGGGCUCUGAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-127-5p | MIMAT0017117 |
| Dasypus novemcinctus | dno-miR-127-5p | MIMAT0047619 |
| Guinea pig | cpo-miR-127-5p | MIMAT0046972 |
| Rabbit | ocu-miR-127-5p | MIMAT0048208 |