Search
Search

| Assay Name: | rno-miR-3569 |
|---|---|
| miRBase Accession Number: | MI0015437 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAGGACAGCAGACUCAGGUC |
| Species: | Rat |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001797, mir-3569 |
Mature miRNA Sequence: | GGAGGACAGCAGACUCAGGUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3569 | MIMAT0017849 |