Search
Search

| Assay Name: | mmu-miR-185-3p |
|---|---|
| miRBase Accession Number: | MI0000227 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGGGCUGGCUUUCCUCUGGU |
| Species: | Mouse, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000202, mir-185 |
Mature miRNA Sequence: | AGGGGCUGGCUUUCCUCUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-185-3p | MIMAT0016996 |
| Cricetulus griseus | cgr-miR-185-3p | MIMAT0023812 |
| Dasypus novemcinctus | dno-miR-185-3p | MIMAT0047706 |
| Guinea pig | cpo-miR-185-3p | MIMAT0047055 |
| Pteropus alecto | pal-miR-185-3p | MIMAT0039841 |
| Rabbit | ocu-miR-185-3p | MIMAT0048292 |
| View More Rows | ||
| Rhesus monkey | mml-miR-185-3p | MIMAT0026831 |