Search
Search

| Assay Name: | dme-miR-278-5p |
|---|---|
| miRBase Accession Number: | MI0000361 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCGGAUGAUGGUUCACAACGACC |
| Species: | Drosophila melanogaster, Drosophila virilis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000155, mir-278 |
Mature miRNA Sequence: | CCGGAUGAUGGUUCACAACGACC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-278-5p | MIMAT0020805 |
| Drosophila virilis | dvi-miR-278-5p | MIMAT0032063 |