Search
Search

| Assay Name: | cgr-miR-17-3p |
|---|---|
| miRBase Accession Number: | MI0020419 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACUGCAGUGCAGGCACUUGUGG |
| Species: | Cricetulus griseus |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCAGUGCAGGCACUUGUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-17-3p | MIMAT0023796 |