Search
Search

| Assay Name: | tur-miR-137-5p |
|---|---|
| miRBase Accession Number: | MI0019398 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGUAUUCCUGGGCUAAUAAC |
| Species: | Tetranychus urticae |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000106, mir-137 |
Mature miRNA Sequence: | GUGUAUUCCUGGGCUAAUAAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tetranychus urticae | tur-miR-137-5p | MIMAT0023098 |