Search
Search

| Assay Name: | hsa-miR-26b-3p |
|---|---|
| miRBase Accession Number: | MI0000084 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCUGUUCUCCAUUACUUGGCU |
| Species: | Human, Pig, Pteropus alecto, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000043, mir-26 |
Mature miRNA Sequence: | CCUGUUCUCCAUUACUUGGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-26b-3p | MIMAT0004500 |
| Pig | ssc-miR-26b-3p | MIMAT0041596 |
| Pteropus alecto | pal-miR-26b-3p | MIMAT0040117 |
| Rhesus monkey | mml-miR-26b-3p | MIMAT0026801 |