Search
Search

| Assay Name: | hsa-miR-552-5p |
|---|---|
| miRBase Accession Number: | MI0003557 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUUUAACCUUUUGCCUGUUGG |
| Species: | Human |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000501, mir-552 |
Mature miRNA Sequence: | GUUUAACCUUUUGCCUGUUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-552-5p | MIMAT0026615 |