Search
Search

| Assay Name: | hsa-miR-7850-5p |
|---|---|
| miRBase Accession Number: | MI0025520 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUUUGGACAUAGUGUGGCUGG |
| Species: | Human |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | GUUUGGACAUAGUGUGGCUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-7850-5p | MIMAT0030425 |