Search
Search

| Assay Name: | mmu-miR-3154 |
|---|---|
| miRBase Accession Number: | MI0030343 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGAAGGGGAGUCGGGAGCGGA |
| Species: | Mouse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0001387, mir-3154 |
Mature miRNA Sequence: | CAGAAGGGGAGUCGGGAGCGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-3154 | MIMAT0035714 |