Search
Search

| Assay Name: | rgl-miR7805-3p |
|---|---|
| miRBase Accession Number: | MI0025458 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUCAUUUACACCAAAUUUGG |
| Species: | Rehmannia glutinosa |
| Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UAUUCAUUUACACCAAAUUUGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rehmannia glutinosa | rgl-miR7805-3p | MIMAT0032248 |