Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12763
          Assay Name: mmu-miR-297a-3p
          miRBase Accession Number: MI0005488
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 10515820 - 10515921 [+] on Build GRCm38
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Species: Mouse
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12763
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000204, mir-297

          Mature miRNA Sequence:

          UAUACAUACACACAUACCCAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-297a-3p MIMAT0004864
          Mouse mmu-miR-297b-3p MIMAT0004827
          Mouse mmu-miR-297c-3p MIMAT0004866

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-297a-3
          Stem-loop Accession # MI0005488
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUGCAUGUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCACACAAACACACAGAAAAUUGA
          Chromosome Location Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004864
          miRBase ID: mmu-miR-297a-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297a-4
          Stem-loop Accession # MI0005489
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUGCAUGUGUGUAUAUGAGUAUACAUAUACAUACACACAUACCCAUACAAGCAUGCACACAAAUACACGCAAAA
          Chromosome Location Chr. 2 - 10517067 - 10517164 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004864
          miRBase ID: mmu-miR-297a-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297b
          Stem-loop Accession # MI0004674
          Stem-loop Sequence
          UCUAUUUGCUUGUGUGUAUAUGUAUGUGUGCAUGAACAUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCAUACAAACACACAGAAAAUGGA
          Chromosome Location Chr. 2 - 10511667 - 10511775 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004827
          miRBase ID: mmu-miR-297b-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10511667 - 10511775 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297c
          Stem-loop Accession # MI0005492
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUACAUGUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCAUACAAACACACAAAAAU
          Chromosome Location Chr. 2 - 10509016 - 10509113 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004866
          miRBase ID: mmu-miR-297c-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10509016 - 10509113 [+] on Build GRCm38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04227838_pri
          TaqMan™ Pri-miRNA Assay : Mm03308471_pri
          TaqMan™ Pri-miRNA Assay : Mm04271926_pri
          TaqMan™ Pri-miRNA Assay : Mm03308455_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12763
          TaqMan™ MicroRNA Assay : 002454
          mirVana® miRNA inhibitor : MH12763
          mirVana® miRNA mimic : MC12763
          TaqMan™ Advanced miRNA Assay : mmu481496_mir


          miRBase Alias:

          Mouse: mmu-miR-297a* (v17)
          Mouse: mmu-miR-297c* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline