Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12763
          Assay Name: mmu-miR-297a-3p
          miRBase Accession Number: MI0005488
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 10515820 - 10515921 [+] on Build GRCm38
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Species: Mouse
          Product Type: mirVana® miRNA mimic
          Assay ID MC12763

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000204, mir-297

          Mature miRNA Sequence:

          UAUACAUACACACAUACCCAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-297a-3p MIMAT0004864
          Mouse mmu-miR-297b-3p MIMAT0004827
          Mouse mmu-miR-297c-3p MIMAT0004866

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-297a-3
          Stem-loop Accession # MI0005488
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUGCAUGUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCACACAAACACACAGAAAAUUGA
          Chromosome Location Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004864
          miRBase ID: mmu-miR-297a-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297a-4
          Stem-loop Accession # MI0005489
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUGCAUGUGUGUAUAUGAGUAUACAUAUACAUACACACAUACCCAUACAAGCAUGCACACAAAUACACGCAAAA
          Chromosome Location Chr. 2 - 10517067 - 10517164 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004864
          miRBase ID: mmu-miR-297a-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10515820 - 10515921 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297b
          Stem-loop Accession # MI0004674
          Stem-loop Sequence
          UCUAUUUGCUUGUGUGUAUAUGUAUGUGUGCAUGAACAUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCAUACAAACACACAGAAAAUGGA
          Chromosome Location Chr. 2 - 10511667 - 10511775 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004827
          miRBase ID: mmu-miR-297b-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10511667 - 10511775 [+] on Build GRCm38

          Stem-loop ID mmu-mir-297c
          Stem-loop Accession # MI0005492
          Stem-loop Sequence
          CUGUGUGUAUAUGUAUGUGUGCAUGUACAUGUGUGUAUAUGAAUAUACAUAUACAUACACACAUACCCAUACAAACAUGCAUACAAACACACAAAAAU
          Chromosome Location Chr. 2 - 10509016 - 10509113 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004866
          miRBase ID: mmu-miR-297c-3p
          Mature miRNA Sequence: UAUACAUACACACAUACCCAUA
          Chromosome Location: Chr. 2 - 10509016 - 10509113 [+] on Build GRCm38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04227838_pri
          TaqMan™ Pri-miRNA Assay : Mm03308471_pri
          TaqMan™ Pri-miRNA Assay : Mm04271926_pri
          TaqMan™ Pri-miRNA Assay : Mm03308455_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12763
          Ambion® Pre-miR™ miRNA Precursor : PM12763
          TaqMan™ MicroRNA Assay : 002454
          mirVana® miRNA inhibitor : MH12763
          TaqMan™ Advanced miRNA Assay : mmu481496_mir


          miRBase Alias:

          Mouse: mmu-miR-297a* (v17)
          Mouse: mmu-miR-297c* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline