Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu478752_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-200a-5p
          Stem-loop Accession Number: MI0000737
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 1167863 - 1167952 [+] on Build GRCh38
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Species: Human, Mouse, Dog, Guinea pig, Pteropus alecto, Rhesus monkey, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu478752_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          CAUCUUACCGGACAGUGCUGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200a-5p MIMAT0001620
          Mouse mmu-miR-200a-5p MIMAT0004619
          Dog cfa-miR-200a MIMAT0009865
          Guinea pig cpo-miR-200a-5p MIMAT0047089
          Pteropus alecto pal-miR-200a-5p MIMAT0039866
          Rhesus monkey mml-miR-200a-5p MIMAT0026842
          View More Rows
          Zebrafish dre-miR-200a-5p MIMAT0031984

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200a
          Stem-loop Accession # MI0000737
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCGC
          Chromosome Location Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001620
          miRBase ID: hsa-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200a
          Stem-loop Accession # MI0000554
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004619
          miRBase ID: mmu-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38

          Dog

          Stem-loop ID cfa-mir-200a
          Stem-loop Accession # MI0010362
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009865
          miRBase ID: cfa-miR-200a
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200a
          Stem-loop Accession # MI0038692
          Stem-loop Sequence
          CAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047089
          miRBase ID: cpo-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-200a
          Stem-loop Accession # MI0032371
          Stem-loop Sequence
          CAUCUUACCGGACAGUGCUGGAUUUCUCGGCUUGACUCUAACACUGUCUGGUAACGAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039866
          miRBase ID: pal-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-200a
          Stem-loop Accession # MI0007663
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026842
          miRBase ID: mml-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-200a
          Stem-loop Accession # MI0002037
          Stem-loop Sequence
          GGCACUUAGCAGCCAUCUUACCGGACAGUGCUGGACUGUAUAACUGUUUUCUAACACUGUCUGGUAACGAUGUUUGUUGGGUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031984
          miRBase ID: dre-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306736_pri
          TaqMan™ Pri-miRNA Assay : Hs03303376_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10250
          Ambion® Pre-miR™ miRNA Precursor : PM10250
          TaqMan™ MicroRNA Assay : 001011
          mirVana® miRNA inhibitor : MH10250
          mirVana® miRNA mimic : MC10250
          TaqMan™ Advanced miRNA Assay : 478752_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline