Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001011
          Assay Name: hsa-miR-200a*
          miRBase Accession Number: MI0000737
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 1167863 - 1167952 [+] on Build GRCh38
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Species: Human, Mouse, Dog, Guinea pig, Pteropus alecto, Rhesus monkey, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001011
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000019, mir-8

          Mature miRNA Sequence:

          CAUCUUACCGGACAGUGCUGGA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-200a-5p MIMAT0001620
          Mouse mmu-miR-200a-5p MIMAT0004619
          Dog cfa-miR-200a MIMAT0009865
          Guinea pig cpo-miR-200a-5p MIMAT0047089
          Pteropus alecto pal-miR-200a-5p MIMAT0039866
          Rhesus monkey mml-miR-200a-5p MIMAT0026842
          View More Rows
          Zebrafish dre-miR-200a-5p MIMAT0031984

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-200a
          Stem-loop Accession # MI0000737
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCGC
          Chromosome Location Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001620
          miRBase ID: hsa-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: Chr. 1 - 1167863 - 1167952 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-200a
          Stem-loop Accession # MI0000554
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004619
          miRBase ID: mmu-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: Chr. 4 - 156054896 - 156054985 [-] on Build GRCm38

          Dog

          Stem-loop ID cfa-mir-200a
          Stem-loop Accession # MI0010362
          Stem-loop Sequence
          CUGGGCCUCUGUGGGCAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009865
          miRBase ID: cfa-miR-200a
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-200a
          Stem-loop Accession # MI0038692
          Stem-loop Sequence
          CAUCUUACCGGACAGUGCUGGAUUUCUUGGCUUGACUCUAACACUGUCUGGUAACGAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047089
          miRBase ID: cpo-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-200a
          Stem-loop Accession # MI0032371
          Stem-loop Sequence
          CAUCUUACCGGACAGUGCUGGAUUUCUCGGCUUGACUCUAACACUGUCUGGUAACGAUGUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039866
          miRBase ID: pal-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-200a
          Stem-loop Accession # MI0007663
          Stem-loop Sequence
          CCGGGCCCCUGUGAGCAUCUUACCGGACAGUGCUGGAUUUCCCAGCUUGACUCUAACACUGUCUGGUAACGAUGUUCAAAGGUGACCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026842
          miRBase ID: mml-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-200a
          Stem-loop Accession # MI0002037
          Stem-loop Sequence
          GGCACUUAGCAGCCAUCUUACCGGACAGUGCUGGACUGUAUAACUGUUUUCUAACACUGUCUGGUAACGAUGUUUGUUGGGUGACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031984
          miRBase ID: dre-miR-200a-5p
          Mature miRNA Sequence: CAUCUUACCGGACAGUGCUGGA
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool B v3.0
          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Cards:

          TaqMan® Rodent MicroRNA B Cards v3.0
          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panels:

          TaqMan® OpenArray® Human MicroRNA Panel
          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306736_pri
          TaqMan™ Pri-miRNA Assay : Hs03303376_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10250
          Ambion® Pre-miR™ miRNA Precursor : PM10250
          mirVana® miRNA inhibitor : MH10250
          mirVana® miRNA mimic : MC10250
          TaqMan™ Advanced miRNA Assay : mmu478752_mir
          TaqMan™ Advanced miRNA Assay : 478752_mir
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel


          miRBase Alias:

          Human: hsa-miR-200a* (v17)
          Mouse: mmu-miR-200a* (v17)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline