Search
Search

| Assay Name: | hsa-miR-200a-5p |
|---|---|
| miRBase Accession Number: | MI0000737 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAUCUUACCGGACAGUGCUGGA |
| Species: | Human, Mouse, Dog, Guinea pig, Pteropus alecto, Rhesus monkey, Zebrafish |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000019, mir-8 |
Mature miRNA Sequence: | CAUCUUACCGGACAGUGCUGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-200a-5p | MIMAT0001620 |
| Mouse | mmu-miR-200a-5p | MIMAT0004619 |
| Dog | cfa-miR-200a | MIMAT0009865 |
| Guinea pig | cpo-miR-200a-5p | MIMAT0047089 |
| Pteropus alecto | pal-miR-200a-5p | MIMAT0039866 |
| Rhesus monkey | mml-miR-200a-5p | MIMAT0026842 |
| View More Rows | ||
| Zebrafish | dre-miR-200a-5p | MIMAT0031984 |