Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu482693_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-188-3p
          Stem-loop Accession Number: MI0000484
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 50003503 - 50003588 [+] on Build GRCh38
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Species: Human, Mouse, Goat, Guinea pig, Horse, Pteropus alecto, Rabbit
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu482693_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000113, mir-188

          Mature miRNA Sequence:

          CUCCCACAUGCAGGGUUUGCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-188-3p MIMAT0004613
          Mouse mmu-miR-188-3p MIMAT0004541
          Goat chi-miR-188-3p MIMAT0036014
          Guinea pig cpo-miR-188-3p MIMAT0047061
          Horse eca-miR-188-3p MIMAT0013199
          Pteropus alecto pal-miR-188-3p MIMAT0040346
          View More Rows
          Rabbit ocu-miR-188-3p MIMAT0048298

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-188
          Stem-loop Accession # MI0000484
          Stem-loop Sequence
          UGCUCCCUCUCUCACAUCCCUUGCAUGGUGGAGGGUGAGCUUUCUGAAAACCCCUCCCACAUGCAGGGUUUGCAGGAUGGCGAGCC
          Chromosome Location Chr. X - 50003503 - 50003588 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004613
          miRBase ID: hsa-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: Chr. X - 50003503 - 50003588 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-188
          Stem-loop Accession # MI0000230
          Stem-loop Sequence
          UCUCACAUCCCUUGCAUGGUGGAGGGUGAGCUCUCUGAAAACCCCUCCCACAUGCAGGGUUUGCAGGA
          Chromosome Location Chr. X - 7247989 - 7248056 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004541
          miRBase ID: mmu-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: Chr. X - 7247989 - 7248056 [-] on Build GRCm38

          Goat

          Stem-loop ID chi-mir-188
          Stem-loop Accession # MI0030659
          Stem-loop Sequence
          UUCCCUGCUCCCUCUCUCACAUCCCUUGCAUGGUGGAGGGUGAGCUUUCUGAAAACCCCUCCCACAUGCAGGGUUUGCAGGAUGGUGAGCCUCGGCUUUUCUUGCUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036014
          miRBase ID: chi-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-188
          Stem-loop Accession # MI0038676
          Stem-loop Sequence
          CAUCCCUUGCAUGGUGGAGGGUGAGCUUUCUGGAAACCCCUCCCACAUGCAGGGUUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047061
          miRBase ID: cpo-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-188
          Stem-loop Accession # MI0012946
          Stem-loop Sequence
          UCUCACAUCCCUUGCAUGGUGGAGGGUGAGCUUUCUGAAAACCCCUCCCACAUGCAGGGUUUGCAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013199
          miRBase ID: eca-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-188
          Stem-loop Accession # MI0032623
          Stem-loop Sequence
          CAUCCCUUGCAUGGUGGAGGGUGCACUUGCUGAAAACCCCUCCCACAUGCAGGGUUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040346
          miRBase ID: pal-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-188
          Stem-loop Accession # MI0039327
          Stem-loop Sequence
          CAUCCCUUGCAUGGUGGAGGGUGAGCUUUAUGAAAACCCCUCCCACAUGCAGGGUUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048298
          miRBase ID: ocu-miR-188-3p
          Mature miRNA Sequence: CUCCCACAUGCAGGGUUUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306463_pri
          TaqMan™ Pri-miRNA Assay : Hs03304216_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12861
          Ambion® Pre-miR™ miRNA Precursor : PM12861
          TaqMan™ MicroRNA Assay : 002106
          mirVana® miRNA inhibitor : MH12861
          mirVana® miRNA mimic : MC12861
          TaqMan™ Advanced miRNA Assay : 477942_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0