Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu483211_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-122b-5p
          Stem-loop Accession Number: MI0040634
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 65248863 - 65248921 [-] on Build GRCm38
          Mature miRNA Sequence: UUUAGUGUGAUAAUGGCGUUUG
          Species: Mouse, Chicken, Monodelphis domestica
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu483211_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UUUAGUGUGAUAAUGGCGUUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-122b-5p MIMAT0049852
          Chicken gga-miR-122b-5p MIMAT0050092
          Monodelphis domestica mdo-miR-122b-5p MIMAT0050214

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-122b
          Stem-loop Accession # MI0040634
          Stem-loop Sequence
          UUUAGUGUGAUAAUGGCGUUUGAUGGUUUGGACACAAACACCAUUGUCACACUCCACAG
          Chromosome Location Chr. 18 - 65248863 - 65248921 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0049852
          miRBase ID: mmu-miR-122b-5p
          Mature miRNA Sequence: UUUAGUGUGAUAAUGGCGUUUG
          Chromosome Location: Chr. 18 - 65248863 - 65248921 [-] on Build GRCm38

          Chicken

          Stem-loop ID gga-mir-122b-1
          Stem-loop Accession # MI0040772
          Stem-loop Sequence
          UUUAGUGUGAUAAUGGCGUUUGAUAGAUUGGACACAAACACCAUUGUCACACUCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050092
          miRBase ID: gga-miR-122b-5p
          Mature miRNA Sequence: UUUAGUGUGAUAAUGGCGUUUG
          Chromosome Location: NA

          Stem-loop ID gga-mir-122b-2
          Stem-loop Accession # MI0040778
          Stem-loop Sequence
          UUUAGUGUGAUAAUGGCGUUUGAUAGAUUGGACACAAACACCAUUGUCACACUCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050092
          miRBase ID: gga-miR-122b-5p
          Mature miRNA Sequence: UUUAGUGUGAUAAUGGCGUUUG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-122b
          Stem-loop Accession # MI0040849
          Stem-loop Sequence
          UUUAGUGUGAUAAUGGCGUUUGAUAGACUGGACACAAACACCAUUGUCACACUCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0050214
          miRBase ID: mdo-miR-122b-5p
          Mature miRNA Sequence: UUUAGUGUGAUAAUGGCGUUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306556_pri
          mirVana® miRNA inhibitor : MH41675
          mirVana® miRNA inhibitor : MH42383
          mirVana® miRNA inhibitor : MH41342
          mirVana® miRNA mimic : MC41675
          mirVana® miRNA mimic : MC42383
          mirVana® miRNA mimic : MC41342

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch