Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178756.4 | NP_848871.1 | 3 | 348 | ||
GenBank mRNA | AK034251.1 | - | 3 | 533 | ||
AK043762.1 | - | 3 | 250 | |||
AK047614.1 | - | 3 | 278 | |||
AK053451.1 | - | 3 | 354 | |||
AK087439.1 | - | 3 | 499 | |||
AK087514.1 | - | 3 | 349 | |||
AK160897.1 | - | 3 | 475 | |||
AY345342.1 | - | 3 | 348 | |||
BC096762.1 | - | 3 | 324 | 30094664 |