Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hs3st3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178870.5 | NP_849201.1 | 1 | 811 | ||
GenBank mRNA | AK051284.1 | - | 1 | 812 | ||
BC094320.1 | - | 1 | 865 | 6400013 |