Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GABRG2 SIRNA Assays ›
          Gene Symbol:
          Gabrg2
          Gene Name:
          gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2
          Gene Aliases:
          GABAA-R, Gabrg-2, gamma2
          Chromosome Location:
          Chr.11: 41910190 - 42000840 on Build GRCm38
          UniGene ID:
          Mm.5309
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 100297

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gabrg2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008073.3 NP_032099.1 1, 2 495
          NM_177408.6 NP_803127.3 1, 2 495
          GenBank mRNA AF233775.1 - 1, 2 111
          AF233776.1 - 1, 2 111
          AF233777.1 - 1, 2 111
          AF233778.1 - 1, 2 111
          AF233779.1 - 1, 2 111
          AF233780.1 - 1, 2 111
          AF233781.1 - 1, 2 111
          AF233782.1 - 1, 2 111
          AF233783.1 - 1, 2 111
          AF233784.1 - 1, 2 111
          AF233785.1 - 1, 2 111
          AF233786.1 - 1, 2 111
          AF233787.1 - 1, 2 111
          AF233788.1 - 1, 2 111
          AF233789.1 - 1, 2 111
          AF233790.1 - 1, 2 111
          AF233791.1 - 1, 2 111
          AF233792.1 - 1, 2 111
          AF233793.1 - 1, 2 111
          AF233794.1 - 1, 2 111
          AF233795.1 - 1, 2 111
          AF233796.1 - 1, 2 111
          AF233797.1 - 1, 2 111
          AF233798.1 - 1, 2 111
          AF233799.1 - 1, 2 111
          AF233800.1 - 1, 2 111
          AF233801.1 - 1, 2 111
          AF233802.1 - 1, 2 111
          AK034338.1 - 1, 2 465
          AK047085.1 - 1, 2 73
          AK049857.1 - 1, 2 138
          AK136889.1 - 1, 2 103
          AK141129.1 - 1, 2 137
          BC031762.1 - 1, 2 313 4504205
          M62374.1 - 1, 2 458
          M86572.1 - 1, 2 287

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          chloride transport
          signal transduction
          gamma-aminobutyric acid signaling pathway
          chemical synaptic transmission
          post-embryonic development
          adult behavior
          synaptic transmission, GABAergic
          cellular response to histamine
          chloride transmembrane transport
          GABA-A receptor activity
          extracellular ligand-gated ion channel activity
          chloride channel activity
          protein binding
          GABA receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline