Search Thermo Fisher Scientific
Gene Symbol: Lama2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008481.2 | NP_032507.2 | 2 | 326 | ||
GenBank mRNA | AK030879.1 | - | 2 | 327 | ||
AK042011.1 | - | 2 | 415 | |||
AK080131.1 | - | 2 | 251 | |||
BB637328.1 | - | 2 | 249 | |||
U12147.1 | - | 2 | 234 |