Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASPH SIRNA Assays ›
Gene Symbol:
Asph
Gene Name:
aspartate-beta-hydroxylase
Gene Aliases:
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37
Chromosome Location:
Chr.4: 9449085 - 9669344 on Build GRCm38
UniGene ID:
Mm.222206
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
100531
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Asph
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001177849.1
NP_001171320.1
2
405
NM_001177850.1
NP_001171321.1
2
405
NM_001177851.1
NP_001171322.1
2
405
NM_001177852.1
NP_001171323.1
2
206
NM_001177853.1
NP_001171324.1
2
206
NM_001177854.1
NP_001171325.1
2
206
NM_001177855.1
NP_001171326.1
2
405
NM_001177856.1
NP_001171327.1
2
405
NM_001290367.1
NP_001277296.1
2
212
NM_023066.3
NP_075553.2
2
405
NM_133723.2
NP_598484.2
2
206
GenBank mRNA
AF221854.1
-
2
181
AF223413.1
-
2
206
AF223414.1
-
2
216
AF289486.1
-
2
327
AF289487.1
-
2
327
AF289488.1
-
2
327
AF289490.1
-
2
163
AF289491.1
-
2
163
AF289492.1
-
2
163
AF302653.2
-
2
105
AF302654.1
-
2
158
AF302655.1
-
2
85
AK009170.1
-
2
131
AK009903.1
-
2
142
AK014674.1
-
2
139
AK017453.1
-
2
138
AK028679.1
-
2
137
AK030293.1
-
2
329
AK030694.1
-
2
140
AK032420.1
-
2
328
AK035735.1
-
2
214
AK049506.1
-
2
329
AK151787.1
-
2
328
AK160984.1
-
2
330
AK165260.1
-
2
351
BC015281.1
-
2
314
BC061098.1
-
2
351
BC128299.1
-
2
134
BC152365.1
-
2
134
BY268464.1
-
2
407
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pattern specification process
negative regulation of cell proliferation
positive regulation of calcium ion transport into cytosol
peptidyl-amino acid modification
regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
regulation of protein stability
activation of store-operated calcium channel activity
response to ATP
limb morphogenesis
peptidyl-aspartic acid hydroxylation
positive regulation of proteolysis
positive regulation of transcription, DNA-templated
regulation of cytosolic calcium ion concentration
oxidation-reduction process
palate development
face morphogenesis
calcium ion transmembrane transport
cellular response to calcium ion
positive regulation of intracellular protein transport
activation of cysteine-type endopeptidase activity
regulation of protein depolymerization
peptide-aspartate beta-dioxygenase activity
calcium ion binding
protein binding
oxidoreductase activity
metal ion binding
dioxygenase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-vvmmd:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0