Search
Search

Exosc8| Gene Symbol: Exosc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027148.3 | NP_081424.3 | 5, 6 | 577 | ||
| GenBank mRNA | AJ505005.1 | - | 5, 6 | 237 | ||
| AK009584.1 | - | 5, 6 | 249 | |||
| AK169719.1 | - | 3, 4 | 180 | |||
| AK220227.1 | - | 5, 6 | 577 | |||
| BC059089.1 | - | 5, 6 | 606 | 6509820 | ||
| BC067250.1 | - | 4, 5 | 220 | |||