Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 2 | 302 | ||
NM_029606.3 | NP_083882.3 | 25 | 2849 | |||
NM_145688.2 | NP_663721.2 | 4 | 524 | |||
GenBank mRNA | AK005611.1 | - | 2 | 303 | ||
AK006504.1 | - | 4 | 510 | |||
BC005723.1 | - | 4 | 336 | |||
BC048677.1 | - | 4 | 496 | |||
BC086808.1 | - | 19 | 2089 | 30632964 | ||
CA464637.1 | - | 4 | 526 |