Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Leprot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175036.4 | NP_778201.1 | 4 | 378 | ||
GenBank mRNA | AJ011565.1 | - | 4 | 351 | ||
AK009569.1 | - | 4 | 379 | |||
AK088848.1 | - | 2 | 195 | |||
AK152008.1 | - | 4 | 445 | |||
AK159489.1 | - | 4 | 444 | |||
AK160453.1 | - | 4 | 392 | |||
AK165821.1 | - | 4 | 437 | |||
AK168597.1 | - | 4 | 414 | |||
BC004744.1 | - | 4 | 389 | 3583760 | ||
BC010289.1 | - | 4 | 299 | 2651399 |