Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nudt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021431.2 | NP_067406.2 | 1 | 760 | ||
GenBank mRNA | AB041576.1 | - | 1 | 506 | ||
AK036077.1 | - | 1 | 91 | |||
AK053310.1 | - | 1 | 540 | |||
AY152853.1 | - | 1 | 506 | |||
BC071274.1 | - | 1 | 564 | 6331160 |
Gene Symbol: Nudt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031664.1 | NP_001026834.1 | 1 | 512 | ||
GenBank mRNA | BC055771.1 | - | 1 | 469 | 6404994 | |
CX209897.1 | - | 1 | 512 |