Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HBB-B1 SIRNA Assays ›
          Gene Symbol:
          Hbb-b1 Hbb-bs Hbb-bt
          Gene Name:
          hemoglobin, beta adult major chain
          hemoglobin, beta adult s chain
          hemoglobin, beta adult t chain
          Gene Aliases:
          AA409645, Hbb1, Hbbt1, Hbbt2, MommeD7, beta1
          Beta-s, Hbbt1, Hbbt2
          Beta-t
          Chromosome Location:
          -
          Chromosome Location:
          Chr.7: 103826523 - 103827928 on Build GRCm38
          Chromosome Location:
          Chr.7: 103812524 - 103813923 on Build GRCm38
          UniGene ID:
          Mm.467412
          Mm.288567
          Mm.467412
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 101818

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hbb-b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278161.1 NP_001265090.1 2 243
          GenBank mRNA AB020013.1 - 2 186
          AB020015.1 - 2 186
          AB364477.1 - 2 189
          Gene Symbol: Hbb-bs
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001201391.1 NP_001188320.1 2 242
          GenBank mRNA AK002258.1 - 2 242
          AK003096.1 - 2 242
          AK003472.1 - 2 242
          AK005442.1 - 2 242
          AK005490.1 - 2 243
          AK010902.1 - 2 242
          AK010980.1 - 2 242
          AK010981.1 - 2 242
          AK010991.1 - 2 242
          AK010993.1 - 2 241
          AK011006.1 - 2 242
          AK011013.1 - 2 242
          AK011016.1 - 2 242
          AK011027.1 - 2 242
          AK011067.1 - 2 242
          AK011069.1 - 2 244
          AK011077.1 - 2 242
          AK011083.1 - 2 242
          AK011102.1 - 2 242
          AK027903.1 - 2 244
          AK027904.1 - 2 242
          AK088149.1 - 2 242
          AK133714.1 - 2 242
          AK147001.1 - 2 244
          AK160629.1 - 2 242
          AK161021.1 - 2 242
          AK165490.1 - 2 242
          AK168562.1 - 2 243
          AK168826.1 - 2 243
          AK168846.1 - 2 242
          Gene Symbol: Hbb-bt
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008220.5 NP_032246.2 2 243
          GenBank mRNA AK002394.1 - 2 242
          AK005496.1 - 2 242
          AK011033.1 - 2 242
          AK011049.1 - 257
          AK011050.1 - 2 243
          AK011052.1 - 2 242
          AK011057.1 - 2 242
          AK011075.1 - 152
          AK012551.1 - 2 242
          AK014364.1 - 2 243
          AK028067.1 - 2 245
          AK167615.1 - 2 242
          AK168412.1 - 2 241
          AK168477.1 - 2 242
          AK168584.1 - 2 242
          AK168819.1 - 2 242

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: M27173.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of eIF2 alpha phosphorylation by heme
          oxygen transport
          hemopoiesis
          erythrocyte development
          oxygen transporter activity
          peroxidase activity
          oxygen binding
          hemoglobin binding
          haptoglobin binding
          hemoglobin alpha binding
          hemoglobin beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0