Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 1, 2 | 354 | ||
GenBank mRNA | AF144702.1 | - | 1, 2 | 356 | ||
AK003054.1 | - | 1, 2 | 160 | |||
AK009041.1 | - | 1, 2 | 148 | |||
AK009152.1 | - | 1, 2 | 164 | |||
AK013230.1 | - | 1, 2 | 152 | |||
AK088984.1 | - | 1, 2 | 164 | |||
BC011436.1 | - | 1, 2 | 167 | 2812494 |