Search Thermo Fisher Scientific
Gene Symbol: Ttbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162864.1 | NP_001156336.1 | 15 | 6774 | ||
GenBank mRNA | AK049158.1 | - | 1 | 1736 | ||
AK173279.1 | - | 2 | 3738 | |||
BC059249.1 | - | 2 | 3978 | |||
BC094327.1 | - | 1 | 2059 |