Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phox2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008888.3 | NP_032914.1 | 1 | 267 | ||
GenBank mRNA | AK052579.2 | - | 1 | 198 | ||
BC079610.1 | - | 1 | 267 | 30360139 | ||
Y14493.1 | - | 1 | 300 |