Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK10 SIRNA Assays ›
          Gene Symbol:
          MAPK10
          Gene Name:
          mitogen-activated protein kinase 10
          Gene Aliases:
          JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
          Chromosome Location:
          Chr.4: 86012296 - 86594117 on Build GRCh38
          UniGene ID:
          Hs.125503
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103378

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318069.1 NP_001304998.1 8, 9 1417
          NM_002753.4 NP_002744.1 8, 9 953
          NM_138980.3 NP_620446.1 8, 9 1444
          NM_138982.3 NP_620448.1 8, 9 1417
          XM_005263129.2 XP_005263186.1 8, 9 1514
          XM_005263130.2 XP_005263187.1 8, 9 910
          XM_005263135.3 XP_005263192.1 8, 9 1311
          XM_006714268.2 XP_006714331.1 8, 9 1184
          XM_011532117.2 XP_011530419.1 8, 9 1017
          XM_011532118.2 XP_011530420.1 8, 9 1110
          XM_011532120.2 XP_011530422.1 8, 9 1080
          XM_011532121.2 XP_011530423.1 8, 9 946
          XM_017008419.1 XP_016863908.1 9, 10 1207
          XM_017008420.1 XP_016863909.1 8, 9 1641
          XM_017008421.1 XP_016863910.1 8, 9 1237
          XM_017008423.1 XP_016863912.1 8, 9 929
          XM_017008424.1 XP_016863913.1 8, 9 1264
          XM_017008425.1 XP_016863914.1 8, 9 937
          XM_017008426.1 XP_016863915.1 8, 9 1265
          XM_017008429.1 XP_016863918.1 8, 9 1641
          XM_017008430.1 XP_016863919.1 8, 9 1019
          XM_017008431.1 XP_016863920.1 8, 9 1237
          XM_017008432.1 XP_016863921.1 8, 9 910
          XM_017008434.1 XP_016863923.1 8, 9 1294
          XM_017008435.1 XP_016863924.1 8, 9 1184
          XM_017008436.1 XP_016863925.1 8, 9 1668
          XM_017008437.1 XP_016863926.1 8, 9 946
          XM_017008438.1 XP_016863927.1 8, 9 937
          XM_017008439.1 XP_016863928.1 8, 9 1265
          XM_017008442.1 XP_016863931.1 10, 11 1659
          XM_017008443.1 XP_016863932.1 10, 11 1310
          XM_017008444.1 XP_016863933.1 10, 11 1632
          XM_017008445.1 XP_016863934.1 7, 8 1141
          XM_017008446.1 XP_016863935.1 5, 6 1833
          XM_017008447.1 XP_016863936.1 7, 8 1279
          XM_017008448.1 XP_016863937.1 7, 8 1252
          XM_017008451.1 XP_016863940.1 7, 8 1078
          XM_017008452.1 XP_016863941.1 7, 8 1279
          GenBank mRNA AK057723.1 - 3, 4 739
          AK124791.1 - 8, 9 1669
          BC022492.1 - 8, 9 1250
          BC035057.1 - 8, 9 1262 5261221
          BC065516.1 - 3, 4 414 5721954
          U07620.1 - 8, 9 954
          U34819.1 - 7, 8 798
          U34820.1 - 7, 8 822

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          protein phosphorylation
          signal transduction
          JNK cascade
          JUN phosphorylation
          response to light stimulus
          Fc-epsilon receptor signaling pathway
          regulation of circadian rhythm
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          JUN kinase activity
          MAP kinase kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0