Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNK2 SIRNA Assays ›
          Gene Symbol:
          TNK2
          Gene Name:
          tyrosine kinase non receptor 2
          Gene Aliases:
          ACK, ACK-1, ACK1, p21cdc42Hs
          Chromosome Location:
          Chr.3: 195863364 - 195909009 on Build GRCh38
          UniGene ID:
          Hs.518513
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103419

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010938.1 NP_001010938.1 13 2307
          NM_001308046.1 NP_001294975.1 12 2320
          NM_005781.4 NP_005772.3 12 2471
          XM_005269268.3 XP_005269325.1 13 2307
          XM_005269270.3 XP_005269327.1 13 2155
          XM_005269274.3 XP_005269331.1 10 1572
          XM_005269275.3 XP_005269332.1 12 2051
          XM_011512317.2 XP_011510619.1 15 2874
          XM_011512318.2 XP_011510620.2 13 2246
          XM_011512320.1 XP_011510622.1 13 2029
          XM_011512321.2 XP_011510623.1 11 1846
          XM_017005508.1 XP_016860997.1 12 2201
          XM_017005509.1 XP_016860998.1 12 2201
          XM_017005510.1 XP_016860999.1 13 2246
          GenBank mRNA AB209338.1 - 8 5733
          AK126712.1 - 1 1368
          AK127455.1 - 4 629
          AK131539.1 - 13 2307
          AK225786.1 - 12 2267
          BC020389.1 - 10 3437
          BC028164.1 - 13 2229
          L13738.2 - 12 2546

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001110147, NM_001289443, NM_016788, AF037260, DQ666696, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          cell surface receptor signaling pathway
          transmembrane receptor protein tyrosine kinase signaling pathway
          small GTPase mediated signal transduction
          phosphorylation
          cell migration
          cell differentiation
          negative regulation of GTPase activity
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          innate immune response
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of clathrin-mediated endocytosis
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          GTPase inhibitor activity
          epidermal growth factor receptor binding
          protein binding
          ATP binding
          identical protein binding
          metal ion binding
          WW domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0