Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GK SIRNA Assays ›
Gene Symbol:
GK
GK3P
Gene Name:
glycerol kinase
glycerol kinase 3 pseudogene
Gene Aliases:
GK1, GKD
GKP3, GKTB
Chromosome Location:
Chr.X: 30653348 - 30731462 on Build GRCh38
Chromosome Location:
Chr.4: 165277792 - 165280023 on Build GRCh38
UniGene ID:
Hs.1466
Hs.654557
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
103668
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000167.5
NP_000158.1
17, 18
1669
NM_001128127.2
NP_001121599.1
17, 18
1669
NM_001205019.1
NP_001191948.1
18, 19
1687
NM_203391.3
NP_976325.1
18, 19
1687
XM_005274488.4
XP_005274545.1
15, 16
1439
XM_006724483.2
XP_006724546.1
18, 19
1706
XM_006724484.2
XP_006724547.1
18, 19
1705
XM_006724485.2
XP_006724548.1
17, 18
1532
XM_006724486.3
XP_006724549.1
16, 17
1517
XM_011545491.2
XP_011543793.1
19, 20
1725
XM_011545492.1
XP_011543794.1
19, 20
1724
XM_011545493.2
XP_011543795.1
17, 18
1407
XM_011545494.2
XP_011543796.1
17, 18
1352
XM_017029409.1
XP_016884898.1
19, 20
1722
XM_017029410.1
XP_016884899.1
16, 17
1501
XM_017029411.1
XP_016884900.1
16, 17
1516
XM_017029412.1
XP_016884901.1
15, 16
1437
GenBank mRNA
AK294304.1
-
18, 20
1493
AK294938.1
-
19, 20
1722
AK308238.1
-
15, 16
1348
AK313215.1
-
17, 18
1605
BC037549.1
-
17, 18
1526
5213716
BC042421.1
-
18, 19
1688
4820945
BC071595.1
-
18, 19
1688
5276056
BX647604.1
-
18, 19
1508
L13943.1
-
17, 18
1556
X68285.1
-
16, 17
1279
X69886.1
-
13, 14
1105
Gene Symbol: GK3P
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_026575.1
1
1868
GenBank mRNA
AK225740.1
-
1
1519
AK292282.1
-
1
1868
BC066960.1
-
1
1524
Back To Top
More Information
Panther Classification:
Molecular Function -
carbohydrate kinase
Gene Ontology Categories:
Function(s)
Process(es)
glycerol metabolic process
triglyceride metabolic process
response to cold
phosphorylation
regulation of fatty acid metabolic process
triglyceride biosynthetic process
glycerol catabolic process
response to drug
glucose homeostasis
response to ethanol
glycerol-3-phosphate biosynthetic process
glycerol kinase activity
protein binding
ATP binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kl7jc:80/100.66.78.35:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0